Learn More
GE Healthcare Mini Q PC 3.2/3 Available on GSA/VA Contract for Federal Government customers only.
Improves resolution, withstands back pressure and minimizes nonspecific adsorption. GE Healthcare Mini Q PC 3.2/3 precision columns are prepacked with MiniBeads for microscale final polishing and analytical ion exchange separations.
Manufacturer:  GE Healthcare 17068601
Catalog No. 45-002-364
Add to cart
Description
Description
Mini Q PC 3.2/3 Precision Column Holder (0.24 ml) enables the use of precision columns with high-performance chromatography systems such as Ä;KTA design.
  • Monodisperse 3µm nonporous beads yield exceptional resolution, speed, reproducibility and durability
  • PC columns are designed for micropurification
  • Nonporous matrix of MiniBeads improves resolution and flow kinetics, better withstands back pressure, and minimizes nonspecific adsorption of the sample
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Mini Q PC 3.2/3
Glass
Monodisperse polystyrene/divinyl benzene particles
3μm, d50v is the median particle size of the cumulative volume distribution.
1 Piece
3.2 mm
0.1 mL/min. - 1.0 mL/min.
4 to 30°C, 20% Ethanol