Learn More
Fisherbrand™ Mini Tube Rotator PROMO
Can be utilized across a broad spectrum of scientific disciplines, including but not limited to: molecular biology, clinical, and biochemistry
Catalog No. 88-861-051PM
Add to cart
Includes:
  • (2) 10/15 mL Paddle
  • (2) 5/7 mL Paddle
  • (2) 1.5/2 mL Paddle
  • (1) 50 mL Paddle
Description
Description
  • Small footprint
  • Digital RPM Display
  • Maintenance free motor with quiet operation
  • Paddles are easily adjustable/interchanged - Each paddle can be adjusted from 0-90 degrees
  • Achieve more effective mixing with “agitation” mode
  • Agitation will occur at every 40 degrees in the rotation cycle
  • Combination of rotation and oscillation provides more effective mixing
  • Extra paddles available
Warranty and Services
2 Years
Specifications
Specifications
10 to 40 rpm
100/240 V
  • 84 x 0.5/0.8 mL tubes
  • 60 x 1.5/2.0 mL tubes
  • 28 x 5/7 mL tubes
  • 28 x 10/15 mL tubes
  • 6 x 50 mL tubes
Accuracy: ±1 rpm
4°C to 40°C
cCSAus
Plug type: US