Learn More
GE Healthcare Whatman™ Mini-UniPrep™ G2 Syringeless Filters Available on GSA/VA Contract for Federal Government customers only.
Combined all-in-one filter for autosampler-supported UHPLC/HPLC analysis.
$449.50 - $4720.00
Specifications
Description Syringeless Filters
Applications Autosampler-supported UHPLC/HPLC analysis
View More Specs
Products 15
Catalog Number Mfr. No. Membrane Pore Size Housing Material Includes Price Quantity & Availability  
Catalog Number Mfr. No. Membrane Pore Size Housing Material Includes Price Quantity & Availability  
09-940-006 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GN203NPEPP
Polypropylene 0.2μ Translucent with standard cap autosampler vial, plunger with attached filter membrane, standard cap Pack of 100 for $449.50
Add to cart
09-940-008 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GN203NPERC
RC 0.2μ Translucent with standard cap autosampler vial, plunger with attached filter membrane, standard cap Pack of 100 for $449.50
Add to cart
09-940-011 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GN503NPEAQU
PVDF 0.2μ Translucent with standard cap autosampler vial, plunger with attached filter membrane, standard cap Pack of 1000 for $4,500.00
Add to cart
09-940-013 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GN503NPEORG
PTFE 0.2μ Translucent with standard cap autosampler vial, plunger with attached filter membrane, standard cap Pack of 1000 for $4,500.00
Add to cart
09-940-014 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GN503NPEPP
Polypropylene 0.2μ Translucent with standard cap autosampler vial, plunger with attached filter membrane, standard cap Pack of 1000 for $4,500.00
Add to cart
09-940-012 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GN503NPENYL
Nylon 0.2μ Translucent with standard cap autosampler vial, plunger with attached filter membrane, standard cap Pack of 1000 for $4,500.00
Add to cart
09-940-015 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GN503NPERC
RC 0.2μ Translucent with standard cap autosampler vial, plunger with attached filter membrane, standard cap Pack of 1000 for $4,500.00
Add to cart
09-940-016 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GN503NPUAQU
PVDF 0.45μ Translucent with standard cap autosampler vial, plunger with attached filter membrane, standard cap Pack of 1000 for $4,500.00
Add to cart
09-940-018 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GN503NPURC
RC 0.45μ Translucent with standard cap autosampler vial, plunger with attached filter membrane, standard cap Pack of 1000 for $4,500.00
Add to cart
09-940-017 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GN503NPUGMF
Glass fiber 0.45μ Translucent with standard cap autosampler vial, plunger with attached filter membrane, standard cap Pack of 1000 for $4,500.00
Add to cart
09-940-020 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GS203NPEPP
Polypropylene 0.2μ Translucent with standard cap autosampler vial, plunger with attached filter membrane, slit septum cap Pack of 100 for $470.50
Add to cart
09-940-047 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GS503NPEAQU
PVDF 0.2μ Translucent with slit septum cap autosampler vial, plunger with attached filter membrane, slit septum cap Pack of 1000 for $4,710.00
Add to cart
09-940-048 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GS503NPENYL
Nylon 0.2μ Translucent with slit septum cap autosampler vial, plunger with attached filter membrane, slit septum cap Pack of 1000 for $4,720.00
Add to cart
09-940-049 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GS503NPUAQU
PVDF 0.45μ Translucent with slit septum cap autosampler vial, plunger with attached filter membrane, slit septum cap Pack of 1000 for $4,710.00
Add to cart
09-940-050 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
GS503NPUORG
PTFE 0.45μ Translucent with slit septum cap autosampler vial, plunger with attached filter membrane, slit septum cap Pack of 1000 for $4,710.00
Add to cart
Description
Description
  • Borosilicate glass chamber with standard or slit septum cap
  • Translucent or amber housing
  • With integrated autosampler vial, plunger with attached filter membrane
  • Holds 0.5mL of unfiltered sample
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Syringeless Filters
Autosampler-supported UHPLC/HPLC analysis