Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Mini-UniPrep™ Syringeless Filter with Slit-septum Cap
All-in-one filtration device for high-throughput HPLC/UHPLC sample preparation.
Supplier: Cytiva US503NPENYL
Description
- Translucent housing helps easy visual inspection
- Available with PES or nylon membrane
- Removes particulates from samples prepared for HPLC/UHPLC analysis
- Column life extended by reducing contamination which cause clogging and back pressure build-up
Specifications
Translucent | |
Nylon | |
HPLC/UHPLC sample preparation, Removes particulates from samples | |
1000 |
Syringeless Filter | |
0.2 μm | |
Polypropylene | |
0.4 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction