Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ Mini-UniPrep™ Syringeless Filter with Slit-septum Cap Available on GSA/VA Contract for Federal Government customers only.

All-in-one filtration device for high-throughput HPLC/UHPLC sample preparation.

$299.00 - $2710.00

Specifications

Capacity 0.4mL
Color Translucent
Description Syringeless Filter
Housing Material Polypropylene
Applications HPLC/UHPLC sample preparation, Removes particulates from samples
View More Specs

Products 2
Catalog Number Mfr. No. Membrane Diameter (Metric) Membrane Price Quantity & Availability  
Catalog Number Mfr. No. Membrane Diameter (Metric) Membrane Price Quantity & Availability  
09-940-053 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
US203NPUPES
PES Pack of 100 for $299.00
Add to cart
 
09-940-054 Available on GSA/VA Contract for Federal Government customers only. View Documents GE Healthcare
US503NPENYL
Nylon Pack of 1000 for $2,710.00
Add to cart
 
Description

Description

  • Translucent housing helps easy visual inspection
  • Available with PES or nylon membrane
  • Removes particulates from samples prepared for HPLC/UHPLC analysis
  • Column life extended by reducing contamination which cause clogging and back pressure build-up

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

0.4mL
Syringeless Filter
HPLC/UHPLC sample preparation, Removes particulates from samples
Translucent
Polypropylene
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit