Learn More
GE Healthcare Whatman™ Mini-UniPrep™ Syringeless Filters: PROMO
Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.
Specifications
Applications Routine analysis; Composite assays; Content uniformity; Protein precipitation; Solubility/dissolubility testing; Light sensitive samples; Ion chromatography
Type Syringeless Filter
View More Specs
Products 3
Catalog Number Mfr. No. Material Pore Size No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. Material Pore Size No. per Pack Price Quantity & Availability  
09-923-101LR View Documents GE Healthcare
UN203NPENYLPM
Nylon (NYL) 0.2μm 100/Pk. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
09-923-29LR View Documents GE Healthcare
UN203NPUNYLPM
Nylon (NYL) 0.45μm 100/Pk. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
09-923-25LR View Documents GE Healthcare
UN203NPUAQUPM
PVDF 0.45μm 100/Pk. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Pre-assembled all-in-one filtration process; eliminates the need for syringe filter, syringe, vial and septum; reduces costs by up to 40%
  • Wide range of membrane choices from 0.2 and 0.45 µm pore sizes to meet specific sample application requirements
  • Compatible with most major autosamplers
  • Fewer consumables required
  • Plunger contains a filtration membrane at one end and a pre-attached cap/septum at the other
  • Can be used either manually or with a compressor unit
  • Processes up to 6 samples at one time, further improving sample processing time and reducing the risk of hand stress
  • Designed to fit into any autosampler accommodating 12 x 32 mm vials
  • OR, septum can be pierced with needle and sample drawn off for manual injection into an analyzer
Specifications
Specifications
Routine analysis; Composite assays; Content uniformity; Protein precipitation; Solubility/dissolubility testing; Light sensitive samples; Ion chromatography
Syringeless Filter