Whatman™ Mini-UniPrep™ Syringeless Filters: Standard Filters: PROMO OFFER
Quick and easy to use—process sample load in one-third the time
Manufacturer: Whatman™ UN203NPEORGPM
 View more versions of this product
Catalog No. 09-923-102PM
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Pore Size 0.2μm
    No. per Pack 100/Pk.
    Type Syringeless Filter
    Material Polytetrafluoroethylene (PTFE)
    Applications Routine analysis; Composite assays; Content uniformity; Protein precipitation; Solubility/dissolubility testing; Light sensitive samples; Ion chromatography
  • Description & Specifications
Related Products
Product Content Feedback