Minigrip™ Enhanced SPECI-GARD™ Biohazard Bags

Transparent colored bags with biohazard logos

Manufacturer: Minigrip LLC SPECPNYY

 View all options for this product

Catalog No. 22-310-006


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Material Polyethylene
    Width (English) 6 in.
    Width (Metric) 15.24cm
    Height (English) 10 in.
    For Use With (Application) Easy-to-open tear line
    Color Yellow
    Height (Metric) 25.4cm
    Includes Press and Close™ liquid tight adhesive closure, perforated opening, 2 pockets, biohazard label
    Product Type Enhanced Speci-gard™ Biohazard Bag

    • Separate document pocket with flap
    • Press and Close™ secure sample closure provides infection prevention and user safety
    • Utilizes “no glove to adhesive” contact capability when sealing bags
    • Easy-to-open tear line
    • Simple instructions for use printed on bags

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit