Minigrip™ Enhanced SPECI-GARD™ Biohazard Bags

Transparent colored bags with biohazard logos

Manufacturer: Minigrip LLCSupplier Diversity Partner SPECPNYP

 View more versions of this product

Catalog No. 22-310-013

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    For Use With (Application) Easy-to-open tear line
    Height (English) 10 in.
    Height (Metric) 25.4cm
    Includes Press and Close™ liquid tight adhesive closure, perforated opening, 2 pockets, biohazard label
    Product Type Enhanced Speci-gard™ Biohazard Bag
    Width (English) 6 in.
    Color Purple
    Material PE (Polyethylene)
    Width (Metric) 15.24cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit