Minigrip™ Lab Guard™ Specimen Bags

Protect from inadvertent exposure to leakage and contamination

Manufacturer: Minigrip LLCSupplier Diversity Partner SBL2X46B

 View more versions of this product

Catalog No. 22-306-325

  Check Availability
Add to Cart

    Specifications

    Zip Close Zipper
    Thickness 1.75mil
    Dimensions (L x W) 6 x 4 in. (15.2 x 10.1cm)
    Length (English) 6 in.
    Length (Metric) 15.2cm
    Width (English) 4 in.
    Width (Metric) 10.1cm
    Material Polyethylene
    For Use With (Application) For transportation of specimens
    Product Style 3-Walled Biohazard Specimen Bag
    Quantity 1000/Cs.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit