Minigrip™ Reloc ZIPPIT™ Clear Non-Biohazard Reclosable Zipper Bags

Economical bags with zipper closure

Manufacturer: Minigrip LLCSupplier Diversity Partner MGZ2P0406

 View more versions of this product

Catalog No. 22-310-022

  Check Availability
Add to Cart

    Specifications

    Product Type Specimen Transport Bag
    Zip Close Zipper
    Color Clear
    Thickness 2 mil
    Dimensions (L x W) 6 x 4 in. (15.2 x 10.2cm)
    Length (English) 6 in.
    Length (Metric) 15.2cm
    Width (English) 4 in.
    Width (Metric) 10.2cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit