Minigrip™ SPECI-GARD™ Specimen Biohazard Transport Bags

Used for specimen transport in hospitals and labs

Manufacturer: Minigrip LLCSupplier Diversity Partner UF95600BBL

 View more versions of this product

Catalog No. 22-310-004

  Check Availability
Add to Cart

    Specifications

    Disposable Yes
    For Use With (Application) Used for specimen transport in hospitals and labs
    Height (English) 10 in.
    Height (Metric) 25.4cm
    Includes Press and Close™ liquid tight adhesive closure and instructions, biohazard label
    Product Type Speci-gard™ Specimen Biohazard Transport Bag
    Width (English) 6 in.
    Color Clear
    Material PE (Polyethylene)
    Width (Metric) 15.24cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit