Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Minigrip™ SPECI-GARD™ Specimen Biohazard Transport Bags
Used for specimen transport in hospitals and labs
Supplier: Minigrip™ UF9566SPBIO
Description
- Features liquid-tight, Press and Close™ adhesive closure system — simply peel away the release liner and press to close
- With printed biohazard logo
- Separate outside pocket holds the requisition slip
- Traditional scored opening
- No glove-to-adhesive contact, ensuring ease of use
- Once closed, the bag stays closed, even in pneumatic tube systems
- Remains closed if air is trapped inside, or if “grabbed,” will not “pop open”
- Available in single-pocket versions when requisitions are not used, or in diagnostic kit packaging
- To open, simply snap at score
- Disposable, eliminating the possibility of cross-contamination caused by reusing bags
- Manufactured with Micro-Tex™ polyethylene for slip-free handling
Order Info
Cat. No. 22-310-001 contains an additional pre-inserted super absorbent tested to absorb up to 60mL of liquid.
Specifications
Biohazard Specimen Transport Bag | |
Single-use | |
Press and Close™ liquid tight adhesive closure, single pocket (2 wall), biohazard label | |
6 in. | |
15.24 cm | |
15.24 cm |
Clear | |
Used for specimen transport in hospitals and labs | |
Polyethylene | |
6 in. | |
6 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction