Learn More
MBL International Corporation MINK1 50 UL DFS Item
Manufacturer:  MBL International Corporation MC7412
Misshapen/NIK-Related Kinase (MINK/MAP4K6) Polyclonal Antibody Size 50 ul Clone Number Polyclonal Host Species Rabbit Conjugate Application IHC
Catalog No. 50-492-25