Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Minocycline hydrochloride, 890-950μg/mg

Click to view available options
Quantity:
500mg
1g
13614-98-7
C23H28ClN3O7
493.941
MFCD00083669
WTJXVDPDEQKTCV-VQAITOIOSA-N
54685925
(4S,4aS,5aR,12aR)-4,7-bis(dimethylamino)-1,10,11,12a-tetrahydroxy-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrochloride
CN(C)C1C2CC3CC4=C(C=CC(=C4C(=C3C(=O)C2(C(=C(C1=O)C(=O)N)O)O)O)O)N(C)C.Cl
13614-98-7
C23H28ClN3O7
493.941
MFCD00083669
WTJXVDPDEQKTCV-VQAITOIOSA-N
54685925
(4S,4aS,5aR,12aR)-4,7-bis(dimethylamino)-1,10,11,12a-tetrahydroxy-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrochloride
CN(C)C1C2CC3CC4=C(C=CC(=C4C(=C3C(=O)C2(C(=C(C1=O)C(=O)N)O)O)O)O)N(C)C.Cl
  • A broad spectrum antibiotic commonly used against bacteria which cause upper respiratory tract infections
Merck Index 14,6202
Solubility Information Soluble in water
Formula Weight 493.94
Physical Form Powder
Boiling Point 813°C
Color Yellow
Melting Point 205°C to 210°C
Quantity 1g
Chemical Name or Material Minocycline hydrochloride, 890 to 950μg/mg

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : QI7630500

storageNote1 : 2° to 8°C