Learn More
Mintie Technologies™ ECU2™ Containment Unit
Indoor mass airborne isolation without binding modifications
Manufacturer:  Mintie Technologies, Inc.Supplier Diversity Partner OA1000V
 View more versions of this product
Catalog No. 19-150-0103
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Two containment units, a 1000V negative air pressure machine and a 5 ft. modular cuff accessory
21 in.
32 in.
75 lb.
45.72cm
HRSA #2-2, CDC, JCAHO
53.34cm
81.28cm
Negative air machine
34.01kg
18 in.
32 x 18 x 21 in. (81.28 x 45.72 x 53.34cm)