Minuteman™ Allergy Vacuum
Four-stage filtration system contains common allergens
Manufacturer: Minuteman International, Inc. 110026
 View all options for this product
Catalog No. 19-063-055
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Product Type Vacuum filter
    Filter Type HEPA
    • Captures common allergens, such as pollen, mold, animal dander, and dust mites that collect in carpeting, draperies, bedding and furniture
    • Traps harmful allergens with progressively finer stages of filtration
    • Four-stage filtration system consists of a disposable paper bag, a washable embossed polyester filter bag, a disposable impact filter and a certified HEPA (High-Efficiency Particulate Air) filter
    • HEPA Filter is 99% efficient at 0.3 microns; pnk impaction filter is 95% efficient at 0.3 microns; embossed polyester filter bag is 99% efficient at 1.5 microns; and disposable paper bag is 99% efficient at 5.0 microns
    • Patented safety interlock system assures user safety and prevents allergens for being redistributed back into the air; vacuum will not operate if HEPA filter is removed
    • 4 gal. (15L) polyethylene tank is noncorrosive and dent-proof with four 2 in. dia. (5cm) casters, patented self sealing intake, and 40 foot cord
  • Description & Specifications
Related Products
Product Content Feedback