Learn More
Minuteman™ Flammable Liquid Recovery Vacuum
For safe recovery of jet fuel and hazardous liquids
Manufacturer:  Minuteman International, Inc. 701024
 View more versions of this product
Catalog No. 18-999-1007
Please call customer service for current pricing. 1-800-766-7000
Requires : Grounded air line hose assembly
Specifications Description & Specifications
Specifications
Special Neoprene 1.5 in. x 25 ft. grounded vacuum hose with Nitrile core, explosion proof operator handle, all Steel dolly with 5 in. dia. 912.7cm conductive wheels for easy transport and an explosion proof tool kit containing an explosion proof hose
7.62m
181 lb.
Air supply hose assembly
25 ft.
82kg