Minuteman™ Vacuum Accessories and Replacement Parts
Manufacturer: Minuteman International, Inc. 110121PKG
 View more versions of this product
Catalog No. 19-163-070
  Check Availability
Add to Cart
    Specifications
    Quantity 12 Pack
    For Use With (Application) Remediation of hazardous molds, spores, bio-haz dust and viral particulate containment
    Product Type Impact filter
    For Use With (Equipment) 6 and 15 gal. Minuteman bio-haz vacuums
  • Description & Specifications
Related Products
Product Content Feedback