Minuteman™ X829 Series HEPA Vacuums

Safe, easy-to-use vacuum for difficult dry and wet cleaning jobs

Manufacturer: Minuteman International, Inc. C8291503

 View more versions of this product

Catalog No. 19-063-032

  Check Availability
Add to Cart

    Specifications

    Capacity (Metric) Dry: 0.23 cu. m.
    Capacity (English) Dry: 0.82 cu. ft.
    Product Type X829 series HEPA vacuum Dry Only
    Filter Type HEPA
    Efficiency 99.99%
    For Use With (Application) Hazardous dusts
    Material Polyethylene
    Voltage 115VAC
    Capacity (English) Tank 15 gal.
    Capacity (Metric) Tank 57L

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit