Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minuteman™ X829 Series HEPA Vacuums

Safe, easy-to-use vacuum for difficult dry and wet cleaning jobs

Manufacturer:  Minuteman International, Inc. C8292006

 View more versions of this product

Catalog No. 19-811-793

Please call customer service for current pricing. 1-800-766-7000


Specifications Description & Specifications

Specifications

110 cfm
Wet: 19 gal. ; Dry; 0.82 cu. ft.
HEPA
Hazardous dusts
240VAC
87 lb.
20 gal.
Wet: 72L; Dry; 0.37 cu.m
X829 series HEPA vacuum Wet/Dry
99.99%
Polyethylene
105 in.
39kg
76L
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit