Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIP-1 delta Recombinant Rabbit Monoclonal Antibody (001)
MIP-1 delta Recombinant Rabbit Monoclonal Antibody (1)
Supplier: Thermo Scientific MA530219
Description

Specifications
MIP-1 delta | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q16663 | |
CCL15 | |
Recombinant Human CCL15/MIP-5/MIP-1 delta protein (Gln22-Ile113). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
001 | |
Unconjugated | |
CCL15 | |
C-C motif chemokine 15; C-C motif chemokine ligand 15; CCL15; CCL15(22-92); CCL15(25-92); CCL15(29-92); chemokine (C-C motif) ligand 15; chemokine CC-2; HCC-2; HCC-2Lkn-1; HMRP-2B; leukotactin 1; Leukotactin-1; LKN1; LKN-1; Macrophage inflammatory protein 5; MIP-1 delta; MIP-1D; MIP5; MIP-5; MIP-5MIP-1d; Mrp-2b; NCC3; NCC-3; NCC-3MIP-1D; NCC3MRP-2B; new CC chemokine 3; SCYA15; SCYA15member 15; SCYL3; small inducible cytokine subfamily A (Cys-Cys), member 15; small-inducible cytokine A15; SY15 | |
Rabbit | |
Protein A | |
RUO | |
6359 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction