Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIP-3-beta Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | MIP-3-beta |
|---|---|
| Concentration | 0.31 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8710
![]()
|
Proteintech Group Inc
133971AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Macrophage Inflammatory Protein-3 beta (MIP-3 beta), also called CCL19, is a chemokine that is expressed in the thymus, lymph nodes and activated bone marrow stromal cells. MIP-3 beta signals through the G protein-coupled receptor, CCR7, to regulate normal lymphocyte recirculation and homing of T and B cells to the lymph nodes. Human MIP-3 beta shows activity on mouse cells.Specifications
| MIP-3-beta | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q99731 | |
| 6363 | |
| MIP-3-beta Fusion Protein Ag4200 | |
| Primary | |
| -20°C | |
| CCL19 |
| 0.31 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Beta chemokine exodus 3, C C motif chemokine 19, CCL19, CK beta 11, CKb11, EBI1 ligand chemokine, ELC, MIP 3 beta, MIP 3b, MIP3B, MIP-3-beta, SCYA19, Small inducible cytokine A19 | |
| CCL19 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title