Learn More
MIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 179771AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-1913
Add to Cart
Description
Description
AQP0 is the most abundant endogenous protein in the plasma membrane of lens fiber cells where it functions not only as a water pore, but it is also involved in fiber-fiber adhesion and is crucial for fiber cell structure and organization. AQP0 contains an additional pore constriction, not seen in any other aquaporin structures, which may be responsible for pore gating. The closed AQP0 pore holds just three water molecules, which are spaced too far apart to form hydrogen bonds with each other. The C-terminal domain of AQP0 undergoes extensive post-translational modification, including many truncations, during lens aging due to the actions of m-calpain, proteases or non-enzymatic mechanisms. These truncation sites may be involved in the development of cataracts.
Specifications
Specifications
MIP
Polyclonal
Unconjugated
MIP
AQP0, Aquaporin 0, LIM1, MIP, MIP26, MP26
Rabbit
Antigen Affinity Chromatography
RUO
17339, 25480, 4284
-20°C
Liquid
Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P09011, P30301, P51180
Mip
MIP Fusion Protein Ag12369
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Recommendations
Recommendations