Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIPEP Mouse anti-Human, Mouse, Rat, DyLight 488, Clone: OTI5E7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272706G
Description
MIPEP Monoclonal specifically detects MIPEP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
MIPEP | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MIPEP | |
Recombinant protein fragment corresponding to amino acids 174-516 of human MIPEP (NP_005923) produced in E.coli. | |
0.1 mL | |
4285.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI5E7 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
HMIP, MIPEC 3.4.24.59, mitochondrial intermediate peptidase | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title