Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIPEP Mouse anti-Human, Rat, Clone: OTI4D11, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526415

Catalog No. PIMA526415


Add to cart

Description

Description

The product of this gene performs the final step in processing a specific class of nuclear-encoded proteins targeted to the mitochondrial matrix or inner membrane. This protein is primarily involved in the maturation of oxidative phosphorylation (OXPHOS)-related proteins. This gene may contribute to the functional effects of frataxin deficiency and the clinical manifestations of Friedreich ataxia.
Specifications

Specifications

MIPEP
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q01992, Q99797
HMIP, MIP
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4285, 81684
Immunohistochemistry (Paraffin), Western Blot
OTI4D11
Unconjugated
MIPEP
Liquid
Mipep
Human recombit protein fragment corresponding to amino acids 174-516 of MIPEP produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit