Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ Label IT™ Plasmid Delivery Controls

Fluorescently labeled plasmid ideal for in vitro and in vivo tracking using direct fluorescent microscopy

$194.00 - $854.00

Specifications

Concentration 0.5mg/mL
Product Type Plasmid Delivery Control
Format Liquid
For Use With (Application) Visualization and tracking of transfected plasmid DNA and in-vitro and in vivo tracking using direct fluorescent microscopy, co-delivery experiments with functional plasmids
Stability 6 Months
View More Specs

Products 4
Catalog Number Mfr. No. Conjugate Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Conjugate Quantity Price Quantity & Availability  
MIR-7904
View Documents
Mirus Bio LLCSupplier Diversity Partner
MIR 7904
Cy™3 10μg
Each for $257.00
Add to cart
 
MIR-7905
View Documents
Mirus Bio LLCSupplier Diversity Partner
MIR 7905
Cy™3 100μg
Each for $854.00
Add to cart
 
MIR-7906
View Documents
Mirus Bio LLCSupplier Diversity Partner
MIR 7906
Fluorescein 10μg
Each for $194.00
Add to cart
 
MIR-7907
View Documents
Mirus Bio LLCSupplier Diversity Partner
MIR 7907
Fluorescein 100μg
Each for $642.00
Add to cart
 
Description

Description

  • Sensitive — Easily visualize transfected cells and assess transfection efficiency using fluorescent microscopy
  • Compatible — Suitable for co-delivery experiments with functional plasmids
  • Ease of use — Supplied as a ready to use 0.5 mg/ml concentration for both in vitro and in vivo tracking studies

Visualization and tracking of transfected plasmid DNA

Specifications

Specifications

0.5mg/mL
Liquid
6 Months
-20°C
Plasmid Delivery Control
Visualization and tracking of transfected plasmid DNA and in-vitro and in vivo tracking using direct fluorescent microscopy, co-delivery experiments with functional plasmids
TE Buffer (10mMTris, pH 7.5, 0.1mM EDTA)
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit