Learn More
Mirus Bio TransIT-EE Hydrodynamic Delivery Solution and Starter Kit
An “Enhanced Expression” in vivo delivery solution that delivers DNA to the livers of laboratory mice via hydrodynamic tail vein injection
$404.00
Specifications
For Use With (Application) | Delivery of DNA expression vectors into live laboratory mice using the hydrodynamic tail vein injection procedure |
---|
Includes: Provided as TransIT-EE Delivery Solution or TransIT-EE Starter Kit including the TransIT-EE Delivery Solution, 10 syringes, needles and alcohol swabs and conical plastic mouse restraint device.
Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity & Availability | ||||
MIR-5340
|
Mirus Bio
MIR 5340 |
1X TransIT™-EE delivery solution, 40 hydrodynamic tail vein injection | 40 Injections |
Each for $404.00
|
|
MIR-5310
|
Mirus Bio
MIR 5310 |
TransIT™-EE delivery solution, 10 syringes, needle and Alcohol swab, conical Plastic Mouse restraint device, 10 hydrodynamic tail vein injection | 10 Injections |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Naked Nucleic Acid Delivery: Achieve increased levels of gene expression than compared to TransIT-QR Delivery Solution, polylysine and PEI methods
- Versatile Platform: Suitable for DNA delivery using methods including hydrodynamic tail vein injection to the mouse strain of your choice
Specifications
Delivery of DNA expression vectors into live laboratory mice using the hydrodynamic tail vein injection procedure |