Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio™ TransIT™-Jukart Transfection Reagent
TransIT™-Jukart Transfection Reagent optimized to achieve 10 – 25% transfection efficiency in Jurkat cells.
Supplier: Mirus Bio™ MIR 2120
Includes: TransIT-Jurkat Transfection Reagent supplied as ready-to-use solution
Description
- TransIT-Jurkat Transfection Reagent supplied as ready-to-use solution.
- Cells can be transfected in serum-containing media, eliminating the need for media changes and improving transfection effeciency.
- Concentration: 10 mg/mL
Specifications
Primary Cells | |
None (Promoterless) | |
10 mg/mL | |
Transfection Reagent | |
Sterile |
Tube | |
Chemical Transfection | |
Gene expression and promoter analysis, optimized for Jurkat cells and other hard-to-transfect cells | |
1 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction