Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-Jukart Transfection Reagent

TransIT™-Jukart Transfection Reagent optimized to achieve 10 – 25% transfection efficiency in Jurkat cells.

Supplier:  Mirus Bio™ MIR 2124

 View more versions of this product

Catalog No. MIR-2124

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Includes: TransIT-Jurkat Transfection Reagent supplied as ready-to-use solution

Description

Description

  • TransIT-Jurkat Transfection Reagent supplied as ready-to-use solution.
  • Cells can be transfected in serum-containing media, eliminating the need for media changes and improving transfection effeciency.
  • Concentration: 10 mg/mL
Specifications

Specifications

Primary Cell, Hard-To-Transfect Cell, Hard-to-Transfect Cells
None (Promoterless)
10 mg/mL
Transfection Reagent
Sterile
Tube
Chemical Transfection
Gene expression and promoter analysis, optimized for Jurkat cells and other hard-to-transfect cells
0.4 mL
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit