Learn More
Mirus Bio™ TransIT™-Oligo Transfection Reagent
Provides efficient transfection of a wide range of oligonucleotides and oligoribonucleotides into a variety of cell types. Transfections do not require media changes and can be performed in serum-containing media.
Supplier: Mirus Bio™ MIR 2164
Description
- Unique Formulation: Maximize transfection performance of oligonucleotides into a wide range of cells
- Low Cellular Toxicity: Maintain cell density and reduce experimental biases
- High Efficiency Delivery: Achieve high transfection efficiency in cells to ensure experimental success
- One milliliter is sufficient to perform up to 500 transfections in 24-well plates
- Tested on a variety of cell lines and oligonucleotides
Specifications
Mammalian | |
None (Promoterless) | |
Provides efficient transfection of a wide range of Oligonucleotides and Oligoribonucleotides into a variety of cell types | |
Sterile |
Tube | |
Chemical Transfection | |
0.4 mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.