Learn More
Mirus Bio TransIT-QR Hydrodynamic Delivery Solution and Starter Kit
A nontoxic, “Quick Recovery” in vivo delivery solution that delivers DNA or siRNA to the livers of laboratory mice using methods including hydrodynamic tail vein injection
$308.00
Specifications
For Use With (Application) | Delivers DNA or siRNA to the livers of laboratory mice using methods including hydrodynamic tail vein injection |
---|
Includes: TransIT-QR Starter Kit:
TransIT-QR Delivery Solution, 10 syringes, needles and alcohol swabs and a conical plastic tube.
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MIR-5240
|
Mirus Bio
MIR 5240 |
40 Injections |
Each for $308.00
|
|
MIR-5210
|
Mirus Bio
MIR 5210 |
10 Injections |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Naked Nucleic Acid Delivery: Achieve higher levels of gene expression than compared to polylysine and PEI methods
- Versatile Platform: For DNA or siRNA delivery using methods including hydrodynamic tail vein injection to the mouse strain of your choice
- Low Toxicity: Minimized loss of cardiac output compared to saline injections, which allows for quick recovery (within minutes) of the mouse post-injection
Specifications
Delivers DNA or siRNA to the livers of laboratory mice using methods including hydrodynamic tail vein injection |