Learn More
Thermo Scientific™ Miscellaneous Microtome Accessories
$975.00
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
14-071-432
View Documents
Thermo Scientific™
640280
Foot Pedal |HM 350S, HM 355S, HM 360
Each for $975.00
Add to cart
40-635-0
View Documents
Thermo Scientific™
406350
Waste Tray |HM 340, HM 355S, HM 360 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Support and expand the functionality of Thermo Scientific™ 200 and 300 series of microtomes with Thermo Scientific Miscellaneous Microtome Accessories.
  • Interlock plug replaces foot pedal connector
  • Storage unit can be removed and used separately; tray for specimens can be cooled by changeable inserts
  • Large-field magnifier attaches to facilitate knife alignment and approach for sectioning; magnification 2.5X