Learn More
Thermo Scientific™ Miscellaneous Microtome Accessories
Support and expand the functionality of Thermo Scientific™ 200 and 300 series of microtomes with Thermo Scientific Miscellaneous Microtome Accessories.
Manufacturer:  Thermo Scientific™ 640280
 View more versions of this product
Catalog No. 14-071-432
  Check Availability
Add to cart
Specifications
Description
Support and expand the functionality of Thermo Scientific™ 200 and 300 series of microtomes with Thermo Scientific Miscellaneous Microtome Accessories.
  • Interlock plug replaces foot pedal connector
  • Storage unit can be removed and used separately; tray for specimens can be cooled by changeable inserts
  • Large-field magnifier attaches to facilitate knife alignment and approach for sectioning; magnification 2.5X
Description & Specifications
Specifications
Foot Pedal|HM 350S, HM 355S, HM 360
Thermo Scientific™ 200/300 Series microtomes
Foot pedal