Learn More
Thermo Scientific™ Miscellaneous Microtome Accessories Available on GSA/VA Contract for Federal Government customers only.
Support and expand the functionality of Thermo Scientific™ 200 and 300 series of microtomes with Thermo Scientific Miscellaneous Microtome Accessories.
Manufacturer:  Thermo Scientific™ 760170
 View more versions of this product
Catalog No. 14-071-23
  Check Availability
Add to cart
Specifications
Description
Support and expand the functionality of Thermo Scientific™ 200 and 300 series of microtomes with Thermo Scientific Miscellaneous Microtome Accessories.
  • Interlock plug replaces foot pedal connector
  • Storage unit can be removed and used separately; tray for specimens can be cooled by changeable inserts
  • Large-field magnifier attaches to facilitate knife alignment and approach for sectioning; magnification 2.5X
Description & Specifications
Specifications
HM 310, HM 650V
Large-field Magnifier