Alfa Aesar™ Misch Metal, 99.8% min. rare earth content, trapezoidal ingot, dryb

Manufacturer: Alfa Aesar™ 66144P2

Quantity 1 lb.

 View more versions of this product

Catalog No. AA66144P2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 62379-61-7
    MDL Number MFCD00144799
    UN Number UN3178
    Chemical Name or Material Misch Metal, Dry

    Specifications

    Quantity 1 lb.
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P280-P370+P378c-P402+P404-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, sand, extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Trapezoid Ingot
    UN Number UN3178
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H228-H260
    Flammable solid.
    In contact with water releases flammable gases which may ignite spontaneously.
    Melting Point ≈845°C
    TSCA TSCA
    MDL Number MFCD00144799
    CAS 62379-61-7
    DOT Information Transport Hazard Class: 4.1; Packaging Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
    Weight ≈500g/ingot
    Color Silver
    Odor Odorless
    Concentration Ce rich (varies) 55 to 70%
    Sensitivity Air & Moisture Sensitive
    Chemical Name or Material Misch Metal, Dry
    Assay ≥99.8%, rare earth content

    GHS H Statement
    H228-H260
    Flammable solid.
    In contact with water releases flammable gases which may ignite spontaneously.

    GHS P Statement
    P210-P231+P232-P280-P370+P378c-P402+P404-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, sand, extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit