Learn More
Mitchell Plastics Balance Shield Guards
Manufacturer:  Mitchell Plastics RP780
 View more versions of this product
Catalog No. 05-723-476
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Protect samples during weighing
  • Enclose balance leaving controls exposed for easy access
  • Hinged lid offers direct access to weighing pan
  • 1 in. hole on top allows for sample dispensing
  • Designed for Mettler and Sartorius balances
Specifications
Specifications
Mettler Balance
6.5 in.
16.51 cm