Learn More
Mitchell Plastics Benchtop Liners
Manufacturer:  Mitchell Plastics SM5
 View more versions of this product
Catalog No. 05-723-413
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
45 x 60cm (17.75 x 23.5 in.)
Rayon, Cellulose and Polyethylene
50 Pack
Used when working with body fluids, tissue samples, pathogens or other biohazardous materials
Biohazard Mat