Mitchell Plastics Biohazard Benchtop Waste Containers

Clear containers facilitate easy and safe segregation of hazardous materials for later disposal

Manufacturer: Mitchell Plastics WB600

 View more versions of this product

Catalog No. 05-723-425

  Check Availability
Add to Cart

Disclaimers:

Dispose contents according to all local, state and federal disposal regulations.


    Specifications

    Color Clear
    Dimensions (L x W x H) 11 x 11 x 11.5 in. (27.9 x 27.9 x 29.21cm)
    For Use With (Application) Waste disposal
    Material Plastic
    Width (English) 11 in.
    Width (Metric) 27.9cm
    Height (Metric) 29.21cm
    Product Type Benchtop Biohazard Waste Container
    Holds 20.32 x 20.32 x 25.4cm Box
    Length (English) 11 in.
    Length (Metric) 27.9cm
    Model Benchtop
    Thickness 4.75mm
    Height (English) 11.5 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit