Learn More
Mitchell Plastics Blood Bag Trays
Keep blood bags conveniently organized
Manufacturer:  Mitchell Plastics BB11002
 View more versions of this product
Catalog No. 22-312-419
Add to cart
Description
Description
  • Fits into most drawer-type refrigerators
  • Made from sturdy clear acrylic, enabling easy visibility of bags and labels
  • Design allows blood bags to be easily stored by type and Rh factor
  • Helps prevent contamination by contact with refrigerator and eliminates cold-spotting
  • May be cleaned with soap, water and gentle disinfectants
Specifications
Specifications
Clear
12.1 cm
5 in.
Blood Bags (2)
12.1 cm
2
12.7 cm
4.75 in.
4.75 x 5 x 5 in. (12.1 x 12.7 x 12.7cm)
12.7 cm
4.75 in.
Acrylic
5 in.
Blood Bag Rack