Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitchell Plastics Refrigerator Storage Box with Lock

Store drugs, reagents, liquids and other contents from unauthorized users with the electronic, touch pad lock or combination locks

$213.50 - $282.50

Specifications

Product Type Storage Box
For Use With (Application) Store Reagents, Liquids and Other Contents from Unauthorized Users with Lockable Storage Boxes
Includes Pull-Out Drawer
Material Acrylic
View More Specs

Products 6
Catalog Number Mfr. No. Color Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Color Dimensions (L x W x H) Price Quantity & Availability  
05-723-471
View Documents
Mitchell Plastics
SBCLCG396
Clear, Neon Green 3.75in. x 9.5in. x 6.625in.
Each for $282.50
Add to cart
 
05-723-473
View Documents
Mitchell Plastics
SBKLW396
Black, White 3.75 x 9.5 x 6.375 in.
Each for $258.50
Add to cart
 
05-723-470
View Documents
Mitchell Plastics
RBCLC4126
Clear 4.25 x 12 x 6 in.
Each for $213.50
Add to cart
 
05-723-472
View Documents
Mitchell Plastics
SBCLW6138
Black, White 6.875 x 13.75 x 8.25 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
05-723-474
View Documents
Mitchell Plastics
SBDLW994
White 9.25 x 9.5 x 4.5 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
05-723-475
View Documents
Mitchell Plastics
SBDLB15154
Black 15.375 x 15.625 x 4.50 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Combination lock provides for use by authorized users without having to locate and share a key
  • Space savings size to full size to accommodate whatever the need
  • Combination locks can be easily programmable
  • Keyed locks available
  • Removable drawer for easy re-filling
  • 3/16 in. thick acrylic provides protection from exposure to beta radiation during transport, storage or general use
  • Solid White Sintra with black drawer available to protect contents from view
  • Top of case has four slots for securing to cabinet shelves or racks in refrigerator
  • Can withstand temperatures as low as -70 ° C
  • Electronic, touch pad digital lock and combination locks
  • Select and change combination easily
  • Full size or space savings size to accommodate whatever the need
  • White, heavy-duty, ABS plastic or black acrylic to conceal contents
  • Security hinged door with security screws
  • 3/16 in. thick acrylic provides protection from exposure to beta radiation during transport, storage or general use
  • Double sided tape also provided to secure to glass shelves
  • Custom sizes are available
Specifications

Specifications

Storage Box
Store Reagents, Liquids and Other Contents from Unauthorized Users with Lockable Storage Boxes
Pull-Out Drawer
Acrylic
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit