Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitchell Plastics Glove Box Holders

Holes and double-sided tape are provided with all glove box holders for mounting to the wall or cabinets

Manufacturer:  Mitchell Plastics MGPCLL2000W

 View more versions of this product

Catalog No. 06-666-811


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Glove Dispenser
21 x 4 x 6.75 in. (53.34 x 10.16 x 17.145cm)
Horizontal or vertical mount
6.75 in.
21.4 in.
4 in.
Epoxy powder coated
White
2 boxes
Double glove box holder
17.145cm
53.34cm
10.16cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit