Learn More
Mitchell Plastics Flexshield™ Benchtop Station and Accessories
Angled shields provides optimum viewing and protective barrier between lab personnel and biohazardous or beta materials
Manufacturer:  Mitchell Plastics RP805
 View more versions of this product
Catalog No. 05-723-405
Add to cart
Description
Description
Specifications
Specifications
Clear
0.75 in.
Benchtop Station and Accessories
Base
16 x 14 in.
Flex shield
Flex shield 06-666-78