Learn More
Mitchell Plastics Flexshield™ Benchtop Station and Accessories
Angled shields provides optimum viewing and protective barrier between lab personnel and biohazardous or beta materials
Manufacturer:  Mitchell Plastics RP862
 View more versions of this product
Catalog No. 05-723-408
Add to cart
Description
Description
Specifications
Specifications
Yellow
Magnetic mount
Magnetic mount
Benchtop Station and Accessories
Accessory Shield