Learn More
Mitchell Plastics Single Blood Bag Holders
Easy-to-use blood bag storage that helps prevent contamination and cold spotting in refrigerator
Manufacturer:  Mitchell Plastics SBBHG12
 View more versions of this product
Catalog No. 22-312-412
Add to cart
Description
Description
The gentle clamp design helps keep bag in place during processing, storage and transport.
  • Keep blood bags upright
  • Clear acrylic enables visibility of bags and labels
  • Compact size allows for easy storage in refrigerator
  • D x W x H: 5 x 11.4 x 10.1cm (2 x 4.5 x 4 in.)
  • May be cleaned with soap, water and gentle disinfectants
  • Available in multiple colors
  • Hold one blood bag
Specifications
Specifications
Neon Green
4.01 in.
2 in.
Acrylic
4.48 in.
2 x 4.48 x 4.01 in. (5.1 x 11.4 x 10.2cm)
10.2 cm
5.1 cm
12 Pack
11.4 cm