Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitchell Plastics Test Tube Dispenser

Conveniently stores and dispenses 12 x 75mm (0.47 x 2.95 in. ) test tubes

Manufacturer:  Mitchell Plastics ML2200

 View more versions of this product

Catalog No. 05-723-453


Add to cart

Description

Description

 

  • Clear, acrylic front panel allows the contents to be viewed
  • Notched opening makes it easy to shake glass fragments from the dispenser while the tubes are in place
  • To load, simply slide the unwrapped test tube box into the dispenser and set it upright

 

Specifications

Specifications

Not Autoclavable
8 x 3.5 x 11.37 in. (20.32 x 8.89 x 28.88cm)
28.88 cm
8 in.
Plastic
8.89 cm
Clear
11.37 in.
Tubes (16 mm x 150 mm)
20.32 cm
3.5 in.
Tube Dispenser
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit