Learn More
Research Products International CorpSupplier Diversity Partner Mitey-Mite Mini-Vortexer with Batteries, Battery Box and AC Adapter DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner 820125
Mitey-Mite is a unique space saving mini vortexer with robust capabilities. Mitey-Mite is battery powered and small enough to fit in crowded spaces or inside fume hoods. Mitey-Mite uses four AA batteries or an AC adapter for power sources. Battery capability is ideal for use in locations where electrical outlets are not available. Features a two position toggle switch for momentary or constant "ON" operation. Designed and built for years of dependable service.
Catalog No. 50-476-592