Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MITF Mouse anti-Human, Clone: SPM290, Abnova™

Mouse monoclonal antibody raised against partial recombinant human MITF.

Manufacturer:  Abnova Corporation MAB13113

Catalog No. 89-934-549


Add to cart

Description

Description

This gene encodes a transcription factor that contains both basic helix-loop-helix and leucine zipper structural features. It regulates the differentiation and development of melanocytes retinal pigment epithelium and is also responsible for pigment cell-specific transcription of the melanogenesis enzyme genes. Heterozygous mutations in the this gene cause auditory-pigmentary syndromes, such as Waardenburg syndrome type 2 and Tietz syndrome. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Specifications

Specifications

MITF
SPM290
Mouse monoclonal antibody raised against partial recombinant human MITF.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
MI/WS2A/bHLHe32
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
MITF
Recombinant protein corresponding to N-terminus of human MITF.
Protein A/G purification
RUO
Primary
4286
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit