Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MITF Rabbit anti-Human, Clone: JF100-01, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266951
Description
MITF Monoclonal antibody specifically detects MITF in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
MITF | |
JF100-01 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
BHLHE32, bHLHe32MI, Class E basic helix-loop-helix protein 32, microphthalmia-associated transcription factor, Waardenburg syndrome, type 2A, WS2A | |
Rabbit | |
IgG | |
100μL | |
Primary | |
4286 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Western Blot 1:1000-1:2000, Flow Cytometry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:100 | |
Purified | |
MITF | |
Recombinant Protein | |
Protein A purified | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title