Learn More
MitMAB, Tocris™
Dynamin inhibitor
Manufacturer:  R&D Systems 4224/50
Quantity 50mg
 View more versions of this product
Catalog No. 42-245-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
336.39
MitMAB
C17H38BrN
Tetradecyltrimethylammonium bromide
Specifications
50mg
>98%
336.39
MitMAB
1119-97-7
C17H38BrN
Tetradecyltrimethylammonium bromide
Safety and Handling
Recommended Storage : Desiccate at Room Temperature